Polyclonal anti-RFP reacts with red fluorescent protein (RFP from Discosoma spp.) and its variants. To minimize cross-reactivity, the antibody has been adsorbed against human, mouse and rat serum proteins. This conjugate is prepared by labeling polyclonal rabbit anti-RFP IgG (H+L) with CF™594 dye. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.
Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.
Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Rabbit
Isotype:
Reactivity: Discosoma spp.