Ethyl-N-(diphenylmethylen)glycinat 98%

ACRO124130050EA 62.1 EUR
ACRO124130050 ACRO124130250
Ethyl-N-(diphenylmethylen)glycinat 98%
N-(Diphenylmethylene)glycine ethyl ester
Formel: C₁₇H₁₇NO₂
Molekulargewicht: 267,32 g/mol
Schmelzpunkt: 51…55 °C
Flammpunkt: >110 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00010590
CAS-Nummer: 69555-14-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 54 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR