1,1'-Bis(di-tert-butylphosphino)ferrocen 98%

Achtung

ACRO429710010EA 75.3 EUR
ACRO429710010 ACRO429712500
1,1'-Bis(di-tert-butylphosphino)ferrocen 98%
1,1'-Bis(di-tert-butylphosphino)ferrocen
Formel: C₂₆H₄₄FeP₂
Molekulargewicht: 474,42 g/mol
Schmelzpunkt: 73…75 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01630818
CAS-Nummer: 84680-95-5

Bestellen

Spezifikation Testergebnisse

Appearance Orange to red Powder
Infrared spectrum Conforms
Melting point 73 to 76 °C
NMR (31P-NMR) consistent with structure
Elemental Analysis 64.18 to 67.47 % C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR