(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate 95%

Lieferant: Thermo Scientific
Achtung

ACRO435820050EA 512 EUR
ACRO435820050
(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate 95%
(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate
Formel: C₁₁H₁₇NO₄S
Schmelzpunkt: 137,3…141 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00040622
CAS-Nummer: 111769-27-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 143 °C
Elemental Analysis 48.91 to 52.99 % C
Elemental Analysis 5.18 to 5.62 % N
Specific optical rotation +3° to +5° (20 °C, 589 nm) (c=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR