3-Ethyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-iumbromid 98%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO437760250EA
67.9
EUR
ACRO437760250
3-Ethyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-iumbromid 98%
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide
Formel:
C₈H₁₄BrNOS Molekulargewicht: 252,16 g/mol Schmelzpunkt: 85…88 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00040549 CAS-Nummer: 54016-70-5 |
Spezifikation Testergebnisse
Appearance | Off-white to orange to light brown Powder or solid |
Infrared spectrum | Conforms |
Titration Argentometric | 97.5 to 102.5 % |
Water | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...