Expanding storage racks, Work2Store®
Lieferant: Heathrow Scientific
Work2Store™
HEATHS120018EA 2010 EUR
HEATHS120018
Expanding storage racks, Work2Store®
Boxen Boxen für Reagenzgläser
Expanding storage rack reduce sample handling time and the need for several racks when working with samples for cold temperature storage. The opening and closing mechanism allows the rack to expand, creating needed room for sample manipulation and then simply collapse the rack back to its compact form and it is ready for the freezer. Temperature range –80 to +121 °C.
  • Instantly creates more working space and helps reduce potential cross contamination
  • Stops opened microfuge lids from interfering with other wells
  • Stackable, autoclavable and with alphanumeric well identifications
  • A low profile clear lid fits either side and locks the rack together for safe handling and transportation
Wells in Work2Store® have a centre depression to stabilise conical tubes and a cryogenic vial lock for one handed cap removal.