Maintenance kit for electrodes, complete, GK ANNEX (Radiometer Analytical)
Lieferant: Hach
HACHS91M001EA 177 EUR
HACHS91M001
Maintenance kit for electrodes, complete, GK ANNEX (Radiometer Analytical)
The GK ANNEX Maintenance Kit contains all that is needed for regular maintenance of combined pH, glass, and reference electrodes with saturated potassium chloride salt bridge.
Kit contains: solutions for normal and intensive cleaning, KCl as saturated solution and crystals for refilling.