Safety collection and transport containers
Lieferant: asecos
MSGA13611EA 35 EUR
MSGA13611
Safety collection and transport containers
Disposal container made of conductive polyethylene.
  • Fully drainable
  • Can be stacked and recycled multiple times
  • Transport approval for packaging group II and III
Zertifizierungen: Type approved in accordance with IMDG, RID, ADRand ICAO-T