Glasröhren
Lieferant: Hecht
HECH42840001EA 8.5 EUR
HECH42840001
Glasröhren
AR-Glass®, clear.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Tube length ~1500 mm