Dispenser for folded hand towels, AQUARIUS* SLIMFOLD
Lieferant: Kimberly-Clark
KIMBU7024EA 157 EUR
KIMBU7024
Dispenser for folded hand towels, AQUARIUS* SLIMFOLD
This white plastic dispenser is ideal for dispensing single sheets of interleaved SCOTT* Slimfold hand towels.
  • Lockable quick-fill dispenser holding up to 250 towels
  • White, high-gloss, easy-clean finish; no dirt or dust traps
  • No need to touch the dispenser, helping reduce contamination and the spread of germs
  • Hassle-free maintenance