Reduced-serum media, MCDB
Lieferant: Biowest
BWSTL1202-500EA 47.6 EUR
BWSTL1202-500
Reduced-serum media, MCDB
These MCDB media were formulated as a reduced serum supplemented medium for the cultivation of human microvascular endothelial cells.
MCDB 131 medium is optimised to support clonal growth of human microvascular endothelial cells, including HUVEC and ECV-304 cells, when supplemented with 0,7% dialysed FBS, 10 ng/ml epidermal growth factor and 1 µg/ml hydrocortisone.