Lanthan(III)nitrat Hexahydrat ≥99%

Lieferant: PanReac AppliChem

APPC122669.1208EA 84.8 EUR
APPC122669.1208
Lanthan(III)nitrat Hexahydrat ≥99%
Lanthan(III)nitrat Hexahydrat
Formel: La(NO₃)₃·6H₂O
Molekulargewicht: 433,01 g/mol
Siedepunkt: 126 °C
Schmelzpunkt: 40 °C
MDL Number: MFCD00149751
CAS-Nummer: 10277-43-7
EINECS: 233-238-0
UN-Nummer: 1477
ADR: 5.1,III

Bestellen

Spezifikation Testergebnisse

Minimum assay (Compl.) 99%
Presentation White Crystals
Solubility Soluble in water 1,580 g/l at 25 °C, Hygroscopic
pH (50 g/l) 3,4
Insoluble matter in H2O 0,005% Max.
Chloride (Cl)  0,005% Max.
Sulphate (SO4 0,002% Max.
Calcium (Ca) 0,005% Max.
Copper (Cu) 0,0005% Max.
Iron (Fe) 0,0005% Max.
Potassium (K) 0,005% Max.
Magnesium (Mg) 0,0005% Max.
Sodium (Na) 0,01% Max.
Nickel (Ni) 0,0005% Max.
Lead (Pb) 0,0005% Max.
Zinc (Zn) 0,0005% Max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR