Probe stand, pHi 105
Lieferant: Beckman Coulter
BECLA58758EA 0 EUR
BECLA58758
Probe stand, pHi 105
Elektrodenhalter
Probe stand for use with either handheld or bench top Φ® series meters.
  • Holds up to four 12 mm Ø electrodes + ATC probe
  • Weighted base provides stability when used separately
  • Includes mounting bracket for direct attachment to Φ® 500 series benchtop meter