Transfection reagent, activated, Penetratin™ 1
Lieferant: MP Biomedicals
Penetratin™ 1
ICNA11PENA0500EA 586 EUR
ICNA11PENA0500
Transfection reagent, activated, Penetratin™ 1
Transfektionsreagenzien
Activated Penetratin™ 1 is a novel tool for peptide and siRNA delivery. It has been successfully used to internalise covalently attached peptides, oligonucleotides, and siRNA.