Total chlorine, test 'n tube vials, DPD reagent
Lieferant: Hach
705-2047EA 106 EUR
HACH2105645
Total chlorine, test 'n tube vials, DPD reagent
For total chlorine determination by the DPD Method. Test 'N Tube Reagent Vials contain pre-measured DPD reagent.
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering