Anti-KLF3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-513EA 537 EUR
PRSI27-513
Anti-KLF3 Rabbit Polyclonal Antibody
Antikörper
KLF3 is a zinc finger transcription factor that is known to function as a potent transcriptional repressor

KLF3 antibody can be used for detection of KLF3 by ELISA at 1:312500. KLF3 antibody can be used for detection of KLF3 by western blot at 1.25 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KLF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR