Meine Sitzung geöffnet lassen?
Endet in 
Ihre Sitzung ist abgelaufen
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Anti-ELF3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
ELF3 is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family. ELF3 contains two putative DNA binding domains: an ETS domain, which is unique in that the 5' half shows relatively weak homology to known ets factors, and an A/T hook domain, found in HMG proteins and various other nuclear factors. In contrast to any known ets factors, ELF3 is expressed exclusively in epithelial cells. ELF3 is expected to be a critical regulator of epithelial cell differentiation. ETS factor ELF3 is a transcriptional regulator of angiopoietin-1 gene regulation in the setting of inflammation. ELF3 overexpressed early during human breast tumorigenesis. ELF3 expression confers a transformed and in vitro metastatic phenotype to otherwise normal MCF-12A cells. ELF3 mediates the expression of TGF-beta RII, and the transcriptional inhibition of ets-related transcription factor could be a one of the mechanisms of colonic carcinogenesis. ELF3 is positively and negatively modulated by other interacting proteins including Ku70, Ku86, p300, and CBP.

ELF3 antibody can be used for detection of ELF3 by ELISA at 1:312500. ELF3 antibody can be used for detection of ELF3 by western blot at 2.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ELF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat




 

BESTELLEN

Erweitert / Kompakt
Größe VE Katalognummer Menge
100 µg 100 µG PRSI27-528
Generate Barcodes Barcode Label Format: 
PDF Katalogseiten erstellen

TECHNISCHE DATEN

Antigen symbol ELF3
Reaktivität Rat, Human
Konjugation Unconjugated
Klonalität Polyclonal
Wirt Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms ESX, ERT, EPR-1, ESE-1
Immunogen A synthetic peptide corresponding a region of human ELF3.
Form Lyophilized
Enter Details

Wählen Sie den Artikel dessen Zertifikat Sie gesucht haben

Chargennummer ist ein Pflichtfeld. Falls Sie diese nicht zur Verfügung haben, tragen Sie "x" für VWR Chemikalien ein.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge