Anti-SNW1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-539EA 589 EUR
PRSI27-539
Anti-SNW1 Rabbit Polyclonal Antibody
Antikörper
SKIIP (Nuclear Protein SkiP, nuclear receptor coactivator NCoA-62, ski-interacting protein) is a member of the SNW gene family, encodes a coactivator that enhances transcription from some Pol II promoters. This coactivator can bind to the ligand-binding domain of the vitamin D receptor and to retinoid receptors to enhance vitamin D-, retinoic acid-, estrogen-, and glucocorticoid-mediated gene expression. It can also interact with poly (A)-binding protein 2 to directly control the expression of muscle-specific genes at the transcriptional level. Finally, the protein may be involved in oncogenesis since it interacts with a region of SKI oncoproteins that is required for transforming activity.

SKIIP antibody can be used for detection of SKIIP by ELISA at 1:62500. SKIIP antibody can be used for detection of SKIIP by western blot at 1.0-5.0 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SNW1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR