Meine Sitzung geöffnet lassen?
Endet in 
Ihre Sitzung ist abgelaufen
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Anti-KIN Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
Kin is a nuclear protein that forms intranuclear foci during proliferation and is redistributed in the nucleoplasm during the cell cycle. Short-wave ultraviolet light provokes the relocalization of the protein, suggesting its participation in the cellular response to DNA damage. Originally selected based on protein-binding with RecA antibodies, the mouse protein presents a limited similarity with a functional domain of the bacterial RecA protein, a characteristic shared by the human ortholog.The protein encoded by this gene is a nuclear protein that forms intranuclear foci during proliferation and is redistributed in the nucleoplasm during the cell cycle. Short-wave ultraviolet light provokes the relocalization of the protein, suggesting its participation in the cellular response to DNA damage. Originally selected based on protein-binding with RecA antibodies, the mouse protein presents a limited similarity with a functional domain of the bacterial RecA protein, a characteristic shared by this human ortholog.

KIN antibody can be used for detection of KIN by ELISA at 1:312500. KIN antibody can be used for detection of KIN by western blot at 2.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Dog




 

BESTELLEN

Erweitert / Kompakt
Größe VE Katalognummer Menge
100 µg 100 µG PRSI27-540
Generate Barcodes Barcode Label Format: 
PDF Katalogseiten erstellen

TECHNISCHE DATEN

Antigen symbol KIN
Antigen name KIN Antigenic determinant of recA Protein homolog
Reaktivität Human, Rat, Dog
Konjugation Unconjugated
Klonalität Polyclonal
Wirt Rabbit
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human KIN.
Form Lyophilized
Enter Details

Wählen Sie den Artikel dessen Zertifikat Sie gesucht haben

Chargennummer ist ein Pflichtfeld. Falls Sie diese nicht zur Verfügung haben, tragen Sie "x" für VWR Chemikalien ein.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge