Meine Sitzung geöffnet lassen?
Endet in 
Ihre Sitzung ist abgelaufen
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Anti-FOXF1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the regulation of pulmonary genes as well as embryonic development.

FOXF1 antibody can be used for detection of FOXF1 by ELISA at 1:62500. FOXF1 antibody can be used for detection of FOXF1 by western blot at 2.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

BESTELLEN

Erweitert / Kompakt
Größe VE Katalognummer Menge
100 µg 100 µG PRSI27-541
Generate Barcodes Barcode Label Format: 
PDF Katalogseiten erstellen

TECHNISCHE DATEN

Antigen symbol FOXF1
Antigen name Forkhead Box F1
Reaktivität Human
Konjugation Unconjugated
Klonalität Polyclonal
Wirt Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms FREAC1, Forkhead drosophila homolog-like 5, FREAC-1, forkhead box protein F1, forkhead-related protein FKHL5, forkhead-related activator 1, FKHL5, ACDMPV, forkhead-related transcription factor 1
Immunogen A synthetic peptide corresponding a region of human FOXF1.
Form Lyophilized
Enter Details

Wählen Sie den Artikel dessen Zertifikat Sie gesucht haben

Chargennummer ist ein Pflichtfeld. Falls Sie diese nicht zur Verfügung haben, tragen Sie "x" für VWR Chemikalien ein.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge