Meine Sitzung geöffnet lassen?
Endet in 
Ihre Sitzung ist abgelaufen
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Anti-PAX3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PAX3 is a member of the paired box (PAX) family of transcription factors. Members of the PAX family typically contain a paired box domain and a paired-type homeodomain. These genes play critical roles during fetal development. Mutations in paired box gene 3 are associated with Waardenburg syndrome, craniofacial-deafness-hand syndrome, and alveolar rhabdomyosarcoma. The translocation t (2;13) (q35;q14), which represents a fusion between PAX3 and the forkhead gene, is a frequent finding in alveolar rhabdomyosarcoma.This gene is a member of the paired box (PAX) family of transcription factors. Members of the PAX family typically contain a paired box domain and a paired-type homeodomain. These genes play critical roles during fetal development. Mutations in paired box gene 3 are associated with Waardenburg syndrome, craniofacial-deafness-hand syndrome, and alveolar rhabdomyosarcoma. The translocation t (2;13) (q35;q14), which represents a fusion between PAX3 and the forkhead gene, is a frequent finding in alveolar rhabdomyosarcoma. Alternative splicing results in transcripts encoding isoforms with different C-termini.This gene is a member of the paired box (PAX) family of transcription factors. Members of the PAX family typically contain a paired box domain and a paired-type homeodomain. These genes play critical roles during fetal development. Mutations in paired box gene 3 are associated with Waardenburg syndrome, craniofacial-deafness-hand syndrome, and alveolar rhabdomyosarcoma. The translocation t (2;13) (q35;q14), which represents a fusion between PAX3 and the forkhead gene, is a frequent finding in alveolar rhabdomyosarcoma. Alternative splicing results in transcripts encoding isoforms with different C-termini.

PAX3 antibody can be used for detection of PAX3 by ELISA at 1:62500. PAX3 antibody can be used for detection of PAX3 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PAX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat




 

BESTELLEN

Erweitert / Kompakt
Größe VE Katalognummer Menge
50 µg 50 µG PRSI27-598
Generate Barcodes Barcode Label Format: 
PDF Katalogseiten erstellen

TECHNISCHE DATEN

Antigen symbol PAX3
Antigen name Paired Box 3
Reaktivität Rat, Human, Mouse
Konjugation Unconjugated
Klonalität Polyclonal
Wirt Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms WS3, WS1, HUP2, paired box protein Pax-3, CDHS, paired domain gene HuP2, paired box homeotic gene 3, paired domain gene 3
Immunogen A synthetic peptide corresponding a region of human PAX3.
Form Lyophilized
Enter Details

Wählen Sie den Artikel dessen Zertifikat Sie gesucht haben

Chargennummer ist ein Pflichtfeld. Falls Sie diese nicht zur Verfügung haben, tragen Sie "x" für VWR Chemikalien ein.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge