Anti-PSMD14 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-600EA 589 EUR
PRSI27-600
Anti-PSMD14 Rabbit Polyclonal Antibody
Antikörper
POH1 (pad one homolog-1) is a component of the 26S proteasome, a multiprotein complex that degrades proteins targeted for destruction by the ubiquitin pathway

PSMD14 antibody can be used for detection of PSMD14 by ELISA at 1:312500. PSMD14 antibody can be used for detection of PSMD14 by western blot at 0.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PSMD14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Drosophila
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR