Anti-SEMA4A Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-813EA 537 EUR
PRSI27-813
Anti-SEMA4A Rabbit Polyclonal Antibody
Antikörper
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.

SEMA4A antibody can be used for detection of SEMA4A by ELISA at 1:312500. SEMA4A antibody can be used for detection of SEMA4A by western blot at 1.25 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SEMA4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR