Anti-IKZF4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-815EA 537 EUR
PRSI27-815
Anti-IKZF4 Rabbit Polyclonal Antibody
Antikörper
Members of the Ikaros (ZNFN1A1; MIM 603023) family of transcription factors, which includes Eos, are expressed in lymphocytes and are implicated in the control of lymphoid development.

ZNFN1A4 antibody can be used for detection of ZNFN1A4 by ELISA at 1:312500. ZNFN1A4 antibody can be used for detection of ZNFN1A4 by western blot at 1.25 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IKZF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR