Anti-ZNF106 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-817EA 589 EUR
PRSI27-817
Anti-ZNF106 Rabbit Polyclonal Antibody
Antikörper
ZFP106 is a new candidate transcription factor

ZFP106 antibody can be used for detection of ZFP106 by ELISA at 1:62500. ZFP106 antibody can be used for detection of ZFP106 by western blot at 0.125 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZNF106
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR