Anti-GZF1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-818EA 589 EUR
PRSI27-818
Anti-GZF1 Rabbit Polyclonal Antibody
Antikörper
ZNF212 belongs to the C2H2-type zinc finger gene family. The zinc finger proteins are involved in gene regulation and development, and are quite conserved throughout evolution.

ZNF336 antibody can be used for detection of ZNF336 by ELISA at 1:12500. ZNF336 antibody can be used for detection of ZNF336 by western blot at 1.0 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GZF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR