Anti-SHOX2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-820EA 537 EUR
PRSI27-820
Anti-SHOX2 Rabbit Polyclonal Antibody
Antikörper
SHOX2 is a member of the homeo box family of genes that encode proteins containing a 60-amino acid residue motif that represents a DNA binding domain. Homeo box genes have been characterized extensively as transcriptional regulators involved in pattern formation in both invertebrate and vertebrate species. Several human genetic disorders are caused by aberrations in human homeo box genes. SHOX is a pseudoautosomal homeo box gene that is thought to be responsible for idiopathic short stature and implicated to play a role in the short stature phenotype of Turner syndrome patients.

SHOX2 antibody can be used for detection of SHOX2 by ELISA at 1:62500. SHOX2 antibody can be used for detection of SHOX2 by western blot at 2.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SHOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR