Anti-ZSCAN18 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI27-823EA 537 EUR
PRSI27-823
Anti-ZSCAN18 Rabbit Polyclonal Antibody
Antikörper
ZNF447 contains 1 SCAN box domain and 2 C2H2-type zinc fingers. It belongs to the kruppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.

ZNF447 antibody can be used for detection of ZNF447 by ELISA at 1:312500. ZNF447 antibody can be used for detection of ZNF447 by western blot at 0.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZSCAN18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR