Anti-OGG1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-994EA 553 EUR
PRSI45-994
Anti-OGG1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 37 kDa band observed in rat spleen lysates (calculated MW of 38.7 kDa according to rat NP_110497.1 and 38.8 kDa according to mouse NP_035087.2). Recommended concentration: 0.1-0.3 μg/mL.

Ogg1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Ogg1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OGG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR