ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 110 kDa band observed in pancreas lysates from rat and mouse (calculated MW of 117 kDa according to NP_858058.1. Recommended concentration: 0.05-0.2 μg/mL. an additional band of unknown identity was also consistently observed at 60 kDa . This band was successfully blocked by incubation with the immunizing peptide.
OGT antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
OGT antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: OGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat