D-Lactose Monohydrat Ph. Eur., Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

SIAL61341-1KGEA 63.1 EUR
SIAL61341-1KG
D-Lactose Monohydrat Ph. Eur., Sigma-Aldrich®
D-Lactose Monohydrat

Lactose is a dissacharide formed by the condensation of one galactose and one glucose molecule. Lactose is the major sugar in the milk of most species, typically present between 2 to 8%. The enzyme lactase hydrolyses lactose to its constituent monosaccharides. In neonates, glucose released via the action of lactase is a major energy source.


Synonyms: β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, lactosum monohydricum, milk sugar.


Zertifizierungen: Tested according to Ph. Eur.

Formel: C₁₂H₂₂O₁₁·1H₂O
Molekulargewicht: 360,32 g/mol
MDL Number: MFCD00150747
CAS-Nummer: 64044-51-5

Bestellen

Spezifikation Testergebnisse

Pharmacopeia Testing conforms to Pharmacopeia

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR