2-(3-Bromphenyl)pyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO457560050EA
315
EUR
ACRO457560050
2-(3-Bromphenyl)pyridin 97%
2-(3-Bromophenyl)pyridine
Formel:
C₁₁H₈BrN Molekulargewicht: 234,0900 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09749846 CAS-Nummer: 4373-60-8 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...