MeOSuc-AAPV-pNA
Lieferant: Enzo Life Sciences
ALEXBML-P213-0005EA 138 EUR
ALEXBML-P213-0005
MeOSuc-AAPV-pNA
Proteine und Peptide
Substrate for neutrophil elastase and neutrophil proteinase 3
Highly sensitive substrate for human and mouse neutrophil elastase (leukocyte elastase) and neutrophil proteinase 3 (PR-3, myeloblastin), but not cathepsin G nor chymotrypsin. This substrate can be used for in vitro assays with purified enzyme, or with biological fluids or conditioned medium. Hydrolysis of the substrate can be detected at 405-410 nm.