Human recombinant IDO,HIS tag
Lieferant: Enzo Life Sciences
ALEXALX2015010050EA 797 EUR
ALEXALX2015010050
Human recombinant IDO,HIS tag
Proteine und Peptide
Produced in E. coli. Mature human IDO (indoleamine 2,3-dioxygenase) (aa 1-403) is fused at the C-terminus to a His-tag.