Gabapentin ≥99% (durch TLC)

Lieferant: Enzo Life Sciences

ALEXBML-CA234-0100EA 478 EUR
ALEXBML-CA234-0100 ALEXBML-CA234-0020
Gabapentin ≥99% (durch TLC)
Gabapentin

Ca2+ channel blocker


Gabapentin is a GABA analog which does not act at GABA receptors but which inhibits L-type calcium channels via binding to the alpha2-delta subunit and neuropathic pain relieving actions. Ligands acting at the alpha2-delta subunit lead to anticonvulsant, anxiolytic and neuropathic pain-relieving actions.

Formel: C₉H₁₇NO₂
Molekulargewicht: 171,24 g/mol
MDL Number: MFCD00865286
CAS-Nummer: 60142-96-3
https://at.vwr.com/stibo/web/std.lang.all/34/78/45403478.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR