[Ala286]-CaM kinase II (281-302)
Lieferant: Enzo Life Sciences
ALEXBML-P202-0500EA 254 EUR
ALEXBML-P202-0500
[Ala286]-CaM kinase II (281-302)
Proteine und Peptide
CaM kinase inhibitor
Sequence is from Ca²⁺/calmodulin-dependent protein kinase II autophosphorylation site within the pseudosubstrate negative regulatory region. Inhibition of CaMKII is as effective as BML-P201 (IC50=2 µM) but kinetics are not complicated by phosphorylation of the peptide by CaMKII itself because of the mutation of Thr286 to Ala.