Z-Leu-Leu-Leu-B(OH)2
Lieferant: Enzo Life Sciences
ALEXBML-PI109-0100EA 371 EUR
ALEXBML-PI109-0100
Z-Leu-Leu-Leu-B(OH)2
Proteine und Peptide
Proteasome inhibitor
A potent cell-permeable proteasome inhibitor (Ki=0,023 nM).