Z-AAF-CMK
Lieferant: Enzo Life Sciences
ALEXBML-PI124-0005EA 105 EUR
ALEXBML-PI124-0005
Z-AAF-CMK
Proteine und Peptide
Negative control for AAF-CMK
Negative control for serine protease inhibitor AAF-CMK which inhibits tripeptidyl peptidase II (TPPII), a giant protease which may substitute for some proteasome functions. Blockage of the N-terminus by the Benzyloxycarbonyl or “Z” group abolishes the activity of the AAF-CMK inhibitor.