Ruxolitinib (free base) ≥99% (durch HPLC)

Lieferant: Enzo Life Sciences

ALEXENZCHM1560010EA 364 EUR
ALEXENZCHM1560010 ALEXENZCHM1560005
Ruxolitinib (free base) ≥99% (durch HPLC)
Ruxolitinib (free base)

JAK inhibitor


Ruxolitinib is a Janus kinase inhibitor with IC50 values of 2.7 and 4.5, respectively, for JAK1 and JAK2.

Formel: C₁₇H₁₈N₆
MDL Number: MFCD12031592
CAS-Nummer: 941678-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR