Dispensing station, electronic, FORTUNA®, OPTIMAT® 3
Lieferant: Poulten & Graf
POUL150.000EA 16646.5 EUR
POUL150.000
Dispensing station, electronic, FORTUNA®, OPTIMAT® 3
This redesigned FORTUNA ® OPTIMAT ® 3 single station automatic programmable dispensing system is suitable for one dosing pump and for continuous operation. Even the dosing of aggressive liquids is possible.
  • Eight pumps available up to 300 ml
  • Precision drive and extremely robust
  • With large colour touch screen display
  • New software for better handling
  • Designed to give years of continuous day-in day-out service
Since there are no metal components involved in the pump, dispensing offers without corrosion or contamination. Mainly involved in the applications such as calibration of volumetric apparatus, extremely accurate dispensing of strong acids, alkalis, high value chemicals in the pharmaceutical and cosmetic industries.
Bestellinformation: Pump and tubings are not included, need to be ordered separately.