Hydroxylammoniumchlorid
|
Achtung
|
Synonyme:
Hydroxylamin Hydrochlorid
, Chlorhydrathydroxylamin
, Hydroxylamin HCl
, Hydroxylamin-Hydrochlorid
MOLE21834412-1KGEA
107
EUR
MOLE21834412-1KG
Hydroxylammoniumchlorid
Hydroxylammoniumchlorid
Formel:
NH₂OH·HCl Molekulargewicht: 69,49 g/mol Siedepunkt: 306 °C (1013 hPa) Schmelzpunkt: 151 °C Dichte: 1,67 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00051089 CAS-Nummer: 5470-11-1 EINECS: 226-798-2 UN-Nummer: 2923 ADR: 8,III Merck Index: 13,04853 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...