4-Methyl-1,10-phenanthrolin

Lieferant: Molekula

MOLEM76544185EA 0 EUR
MOLEM76544185
4-Methyl-1,10-phenanthrolin
4-Methyl-1,10-phenanthroline
Formel: C₁₃H₁₀N₂
MDL Number: MFCD00004978
CAS-Nummer: 31301-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR