2'-Hydroxyacetanilid

Lieferant: Molekula

Synonyme: Ethansäure(2-hydroxyphenyl)amid2-AcetamidophenolN-Acetyl-2-aminophenolN-(2-Hydroxyphenyl)acetamid

MOLE12274085-100GEA 171 EUR
MOLE12274085-100G
2'-Hydroxyacetanilid
2'-Hydroxyacetanilid
Formel: C₈H₉NO₂
Molekulargewicht: 151,16 g/mol
MDL Number: MFCD00002181
CAS-Nummer: 614-80-2
EINECS: 210-396-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR