4-Methoxyzimtsäure

Lieferant: Molekula
Achtung

MOLE40437235-100GEA 0 EUR
MOLE40437235-100G
4-Methoxyzimtsäure
4-Methoxyzimtsäure
Formel: C₁₀H₁₀O₃
Molekulargewicht: 178,18 g/mol
MDL Number: MFCD00004398
CAS-Nummer: 830-09-1
EINECS: 212-594-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR