5-Bromisatin

Lieferant: Molekula

Synonyme: 5-Brom-1H-indol-2,3-dion5-Brom-2,3-indolindion

MOLEM63936113EA 0 EUR
MOLEM63936113
5-Bromisatin
5-Bromisatin
Formel: C₈H₄BrNO₂
Molekulargewicht: 226,03 g/mol
MDL Number: MFCD00149345
CAS-Nummer: 87-48-9
EINECS: 201-747-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR