5-Fluorisatin
Lieferant: Molekula
|
Achtung
|
Synonyme:
5-Fluor-2,3-indolindion
, 5-Fluor-2,3-dioxoindolin
, 5-Fluor-1H-indol-2,3-dion
, 5-Fluorindolin-2,3-dion
MOLEM11960052EA
0
EUR
MOLEM11960052
5-Fluorisatin
5-Fluorisatin
Formel:
C₈H₄FNO₂ Molekulargewicht: 165,12 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00022795 CAS-Nummer: 443-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...