2-Methoxyphenylboronsäure
Lieferant: Molekula
Synonyme:
2-Methoxybenzolboronsäure
, Dihydroxy(2-methoxyphenyl)boran
, (2-Methoxyphenyl)metaborsäure
MOLE24085771-1GEA
0
EUR
MOLE24085771-1G
2-Methoxyphenylboronsäure
2-Methoxyphenylboronsäure
Formel:
C₇H₉BO₃ Molekulargewicht: 151,96 g/mol Schmelzpunkt: 165 °C |
MDL Number:
MFCD00236047 CAS-Nummer: 5720-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...