4-Amino-2-chlorphenol

Lieferant: Molekula

Synonyme: 3-Chlor-4-hydroxyanilin

MOLEM65822552EA 0 EUR
MOLEM65822552
4-Amino-2-chlorphenol
4-Amino-2-chlorphenol
Formel: C₆H₆ClNO
Molekulargewicht: 143,57 g/mol
MDL Number: MFCD00128891
CAS-Nummer: 3964-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR