3'-Nitropropiophenon

Lieferant: Molekula

Synonyme: m-Nitropropiophenon

MOLEM76049489EA 0 EUR
MOLEM76049489
3'-Nitropropiophenon
3'-Nitropropiophenon
Formel: C₉H₉NO₃
Molekulargewicht: 179,18 g/mol
MDL Number: MFCD00051516
CAS-Nummer: 17408-16-1
EINECS: 241-435-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR