2,2',4,5,5'-Pentachlorbiphenyl (PCB Nr. 101)
Lieferant: EHRENSTORFER GMBH
EHERC20010100EA
65.8
EUR
EHERC20010100
2,2',4,5,5'-Pentachlorbiphenyl (PCB Nr. 101)
2,2',4,5,5'-Pentachlorbiphenyl (PCB Nr. 101)
Formel:
C₁₂H₅Cl₅ Molekulargewicht: 326,44 g/mol Schmelzpunkt: 76,5 °C Dichte: 1,28 g/cm³ (20 °C) |
CAS-Nummer:
37680-73-2 UN-Nummer: 2315 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...